3-(Bromomethyl)-4-fluorobenzoic acid

Lieferant: Apollo Scientific
Gefahr

PC500460-1G PC500460-250MG PC500460-5G
APOSPC500460-1GEA 708 EUR
APOSPC500460-1G APOSPC500460-250MG APOSPC500460-5G
3-(Bromomethyl)-4-fluorobenzoic acid
3-(Bromomethyl)-4-fluorobenzoic acid
Formel: C₈H₆BrFO₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD18378684
CAS-Nummer: 89540-20-5
UN-Nummer: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR