6-Fluoro-2-hydroxy-3-nitrobenzonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Cyano-3-fluoro-6-nitrophenol, 3-Cyano-4-fluoro-2-hydroxynitrobenzene

PC500289-1G PC500289-250MG
APOSPC500289-1GEA 319 EUR
APOSPC500289-1G APOSPC500289-250MG
6-Fluoro-2-hydroxy-3-nitrobenzonitrile
6-Fluoro-2-hydroxy-3-nitrobenzonitrile
Formel: C₇H₃FN₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD27975499
CAS-Nummer: 1820648-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR