5-Bromo-4-fluoro-2-nitrophenylacetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-(carboxymethyl)-5-fluoronitrobenzene, (5-Bromo-4-fluoro-2-nitrophenyl)acetic acid

PC500256-1G PC500256-250MG
APOSPC500256-1GEA 464 EUR
APOSPC500256-1G APOSPC500256-250MG
5-Bromo-4-fluoro-2-nitrophenylacetic acid
5-Bromo-4-fluoro-2-nitrophenylacetic acid
Formel: C₈H₅BrFNO₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD27429289
CAS-Nummer: 1598963-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR