5-Bromo-6-fluoro-2-oxindole, tech

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1,3-dihydro-6-fluoro-2H-indol-2-one, 5-Bromo-6-fluoroindolin-2-one

PC500252-1G PC500252-250MG
APOSPC500252-1GEA 823 EUR
APOSPC500252-1G APOSPC500252-250MG
5-Bromo-6-fluoro-2-oxindole, tech
5-Bromo-6-fluoro-2-oxindole
Formel: C₈H₅BrFNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD26086914
CAS-Nummer: 944805-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR