5-Bromo-4-fluoro-2-nitrotoluene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-5-fluoro-2-methylnitrobenzene, 1-Bromo-2-fluoro-5-methyl-4-nitrobenzene
PC500249-1G
PC500249-5G
PC500249-10G
APOSPC500249-1GEA
23.2
EUR
APOSPC500249-1G
APOSPC500249-5G
APOSPC500249-10G
5-Bromo-4-fluoro-2-nitrotoluene
5-Bromo-4-fluoro-2-nitrotoluene
Formel:
C₇H₅BrFNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07777174 CAS-Nummer: 948294-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...