4-Chloro-2-fluoro-5-methoxyaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-chloro-4-fluoroanisole

PC500247-1G PC500247-250MG PC500247-5G
APOSPC500247-1GEA 219 EUR
APOSPC500247-1G APOSPC500247-250MG APOSPC500247-5G
4-Chloro-2-fluoro-5-methoxyaniline
4-Chloro-2-fluoro-5-methoxyaniline
Formel: C₇H₇ClFNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD17169196
CAS-Nummer: 91167-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR