2-Chloro-4-fluoro-5-iodophenol

Lieferant: Apollo Scientific
Achtung

PC500243-1G PC500243-250MG PC500243-5G
APOSPC500243-1GEA 136 EUR
APOSPC500243-1G APOSPC500243-250MG APOSPC500243-5G
2-Chloro-4-fluoro-5-iodophenol
2-Chloro-4-fluoro-5-iodophenol
Formel: C₆H₃ClFIO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22628048
CAS-Nummer: 148254-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR