4-Bromo-2-fluoro-5-nitroanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-5-fluoro-4-methoxy-2-nitrobenzene

PC500241-1G PC500241-250MG
APOSPC500241-1GEA 50.9 EUR
APOSPC500241-1G APOSPC500241-250MG
4-Bromo-2-fluoro-5-nitroanisole
4-Bromo-2-fluoro-5-nitroanisole
Formel: C₇H₅BrFNO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11100974
CAS-Nummer: 661463-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR