2',6'-Difluoro-3'-(trifluoromethyl)acetophenone

Lieferant: Apollo Scientific
Achtung

PC500227-1G PC500227-250MG PC500227-5G
APOSPC500227-1GEA 58 EUR
APOSPC500227-1G APOSPC500227-250MG APOSPC500227-5G
2',6'-Difluoro-3'-(trifluoromethyl)acetophenone
2',6'-Difluoro-3'-(trifluoromethyl)acetophenone
Formel: C₉H₅F₅O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD27937175
CAS-Nummer: 1202679-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR