6-Bromo-7-fluorobenzo[b]furan-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC500193-1G
PC500193-250MG
APOSPC500193-1GEA
300
EUR
APOSPC500193-1G
APOSPC500193-250MG
6-Bromo-7-fluorobenzo[b]furan-2-carboxylic acid
6-Bromo-7-fluorobenzo[b]furan-2-carboxylic acid
Formel:
C₉H₄BrFO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD27664866 CAS-Nummer: 1823358-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...