2-Bromo-6-(trifluoromethyl)phenol 95+%

Lieferant: Apollo Scientific

PC500167-1G PC500167-250MG
APOSPC500167-1GEA 260 EUR
APOSPC500167-1G APOSPC500167-250MG
2-Bromo-6-(trifluoromethyl)phenol 95+%
2-Bromo-6-(trifluoromethyl)phenol
Formel: C₇H₄BrF₃O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13194490
CAS-Nummer: 2844-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR