3-Bromo-5-fluoroisatoic anhydride

Lieferant: Apollo Scientific
Achtung

PC500018-1G PC500018-250MG PC500018-5G
APOSPC500018-1GEA 905 EUR
APOSPC500018-1G APOSPC500018-250MG APOSPC500018-5G
3-Bromo-5-fluoroisatoic anhydride
3-Bromo-5-fluoroisatoic anhydride
Formel: C₈H₃BrFNO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18269603
CAS-Nummer: 1343359-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR