2-Bromo-5-fluoro-4-nitrophenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-fluoro-4-hydroxynitrobenzene
PC49642-1G
PC49642-250MG
PC49642-5G
APOSPC49642-1GEA
347
EUR
APOSPC49642-1G
APOSPC49642-250MG
APOSPC49642-5G
2-Bromo-5-fluoro-4-nitrophenol
2-Bromo-5-fluoro-4-nitrophenol
Formel:
C₆H₃BrFNO₃ Schmelzpunkt: 110…115 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22034975 CAS-Nummer: 1369139-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...