3-Bromo-2-fluoro-5-nitrophenyl methyl sulphone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-fluoro-3-(methylsulphonyl)-5-nitrobenzene, 3-Bromo-4-fluoro-5-(methylsulphonyl)nitrobenzene
PC49612-1G
PC49612-250MG
PC49612-5G
APOSPC49612-1GEA
418
EUR
APOSPC49612-1G
APOSPC49612-250MG
APOSPC49612-5G
3-Bromo-2-fluoro-5-nitrophenyl methyl sulphone 95%
3-Bromo-2-fluoro-5-nitrophenyl methyl sulphone
Formel:
C₇H₅BrFNO₄S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD25541887 CAS-Nummer: 1445995-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...