2-Bromo-4-chloro-6-fluorobenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-2-(bromomethyl)-5-chloro-3-fluorobenzene
PC49476-1G
PC49476-250MG
PC49476-5G
APOSPC49476-1GEA
567
EUR
APOSPC49476-1G
APOSPC49476-250MG
APOSPC49476-5G
2-Bromo-4-chloro-6-fluorobenzyl bromide 95%
2-Bromo-4-chloro-6-fluorobenzyl bromide
Formel:
C₇H₄Br₂ClF Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD31558079 CAS-Nummer: 2090554-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...