2-Bromo-6-fluoro-4-(trifluoromethyl)benzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC49435-1G PC49435-250MG PC49435-5G
APOSPC49435-1GEA 24.8 EUR
APOSPC49435-1G APOSPC49435-250MG APOSPC49435-5G
2-Bromo-6-fluoro-4-(trifluoromethyl)benzaldehyde 95%
2-Bromo-6-fluoro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₃BrF₄O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31665800
CAS-Nummer: 2092468-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR