4-Cyclopropyl-3-fluoroaniline
Lieferant: Apollo Scientific
|
Achtung
|
PC49420-1G
PC49420-250MG
PC49420-5G
APOSPC49420-1GEA
493
EUR
APOSPC49420-1G
APOSPC49420-250MG
APOSPC49420-5G
4-Cyclopropyl-3-fluoroaniline
4-Cyclopropyl-3-fluoroaniline
Formel:
C₉H₁₀FN Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD14585553 CAS-Nummer: 1208083-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...