3-Fluoro-2-iodobenzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC49374-1G PC49374-250MG PC49374-5G
APOSPC49374-1GEA 62.5 EUR
APOSPC49374-1G APOSPC49374-250MG APOSPC49374-5G
3-Fluoro-2-iodobenzaldehyde 97%
3-Fluoro-2-iodobenzaldehyde
Formel: C₇H₄FIO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07782054
CAS-Nummer: 905808-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR