5-(Pentafluorothio)-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1H-Benzimidazol-5-yl)sulphur pentafluoride, 5-(Pentafluorosulphanyl)-1H-benzimidazole
PC49306-1G
PC49306-250MG
APOSPC49306-1GEA
279
EUR
APOSPC49306-1G
APOSPC49306-250MG
5-(Pentafluorothio)-1H-benzimidazole
5-(Pentafluorothio)-1H-benzimidazole
Formel:
C₇H₅F₅N₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22370117 CAS-Nummer: 663179-60-0 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...