3-Bromo-4-(trifluoromethoxy)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

PC49293-1G PC49293-250MG
APOSPC49293-1GEA 186 EUR
APOSPC49293-1G APOSPC49293-250MG
3-Bromo-4-(trifluoromethoxy)benzeneboronic acid
3-Bromo-4-(trifluoromethoxy)benzeneboronic acid
Formel: C₇H₅BBrF₃O₃
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD22580724
CAS-Nummer: 2304634-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR