2-Bromo-6-fluorophenylacetylene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-ethynyl-3-fluorobenzene
PC49284-1G
PC49284-250MG
APOSPC49284-1GEA
406
EUR
APOSPC49284-1G
APOSPC49284-250MG
2-Bromo-6-fluorophenylacetylene
2-Bromo-6-fluorophenylacetylene
Formel:
C₈H₄BrF Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD22460481 CAS-Nummer: 1401670-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...