5-Bromo-2-fluorophenylacetylene

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-ethynyl-1-fluorobenzene

PC49283-1G PC49283-250MG PC49283-5G
APOSPC49283-1GEA 92.8 EUR
APOSPC49283-1G APOSPC49283-250MG APOSPC49283-5G
5-Bromo-2-fluorophenylacetylene
5-Bromo-2-fluorophenylacetylene
Formel: C₈H₄BrF
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08703533
CAS-Nummer: 657427-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR