2-Fluoro-4-(hydroxymethyl)phenol
Lieferant: Apollo Scientific
|
Achtung
|
PC48800-1G
PC48800-250MG
APOSPC48800-1GEA
497
EUR
APOSPC48800-1G
APOSPC48800-250MG
2-Fluoro-4-(hydroxymethyl)phenol
2-Fluoro-4-(hydroxymethyl)phenol
Formel:
C₇H₇FO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06202638 CAS-Nummer: 96740-93-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...