4,6-Dichloro-2-fluoro-3-methoxybenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC48796-1G
PC48796-250MG
PC48796-5G
APOSPC48796-1GEA
260
EUR
APOSPC48796-1G
APOSPC48796-250MG
APOSPC48796-5G
4,6-Dichloro-2-fluoro-3-methoxybenzoic acid
4,6-Dichloro-2-fluoro-3-methoxybenzoic acid
Formel:
C₈H₅Cl₂FO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD31742296 CAS-Nummer: 2365420-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...