3-Bromo-5-fluoro-2-methoxybenzaldehyde 97%
Lieferant: Apollo Scientific
PC48650-1G
PC48650-5G
APOSPC48650-1GEA
87
EUR
APOSPC48650-1G
APOSPC48650-5G
3-Bromo-5-fluoro-2-methoxybenzaldehyde 97%
3-Bromo-5-fluoro-2-methoxybenzaldehyde
Formel:
C₈H₆BrFO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11520672 CAS-Nummer: 1009093-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...