3-Bromo-2-fluoro-5-nitrobenzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC48646-1G PC48646-250MG PC48646-5G
APOSPC48646-1GEA 433 EUR
APOSPC48646-1G APOSPC48646-250MG APOSPC48646-5G
3-Bromo-2-fluoro-5-nitrobenzoic acid 97%
3-Bromo-2-fluoro-5-nitrobenzoic acid
Formel: C₇H₃BrFNO₄
Molekulargewicht: 264.005 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD24107180
CAS-Nummer: 1526564-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR