(S)-2-Chloro-1-(4-fluorophenyl)ethanol 98%

Lieferant: Apollo Scientific
Gefahr

PC48643-1G PC48643-250MG
APOSPC48643-1GEA 524 EUR
APOSPC48643-1G APOSPC48643-250MG
(S)-2-Chloro-1-(4-fluorophenyl)ethanol 98%
(S)-2-Chloro-1-(4-fluorophenyl)ethanol
Formel: C₈H₈ClFO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07776653
CAS-Nummer: 126534-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR