3-Bromo-6-chloro-2-fluoroiodobenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-chloro-2-fluoro-3-iodobenzene

PC48600-1G PC48600-250MG
APOSPC48600-1GEA 95.7 EUR
APOSPC48600-1G APOSPC48600-250MG
3-Bromo-6-chloro-2-fluoroiodobenzene
3-Bromo-6-chloro-2-fluoroiodobenzene
Formel: C₆H₂BrClFI
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28122825
CAS-Nummer: 1820641-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR