6-Bromo-3-chloro-2-fluoroiodobenzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-4-chloro-3-fluoro-2-iodobenzene
PC48574-1G
PC48574-250MG
APOSPC48574-1GEA
145
EUR
APOSPC48574-1G
APOSPC48574-250MG
6-Bromo-3-chloro-2-fluoroiodobenzene
6-Bromo-3-chloro-2-fluoroiodobenzene
Formel:
C₆H₂BrClFI Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28103322 CAS-Nummer: 1820674-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...