5-Bromo-4-fluoro-2-methylphenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-5-fluoro-2-hydroxytoluene, 5-Bromo-4-fluoro-o-cresol, 5-Bromo-4-fluoro-2-methylphenol
PC48458-1G
PC48458-250MG
APOSPC48458-1GEA
290
EUR
APOSPC48458-1G
APOSPC48458-250MG
5-Bromo-4-fluoro-2-methylphenol
5-Bromo-4-fluoro-2-methylphenol
Formel:
C₇H₆BrFO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28096618 CAS-Nummer: 1701766-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...