4-(Difluoromethyl)benzyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Bromomethyl)benzal fluoride, 1-(Bromomethyl)-4-(difluoromethyl)benzene

PC48439-1G PC48439-250MG
APOSPC48439-1GEA 337 EUR
APOSPC48439-1G APOSPC48439-250MG
4-(Difluoromethyl)benzyl bromide
4-(Difluoromethyl)benzyl bromide
Formel: C₈H₇BrF₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD18791603
CAS-Nummer: 873373-34-3
UN-Nummer: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR