2,2-Difluoroethyl ethyl carbonate
Lieferant: Apollo Scientific
|
Achtung
|
PC48383-1G
PC48383-5G
APOSPC48383-1GEA
198
EUR
APOSPC48383-1G
APOSPC48383-5G
2,2-Difluoroethyl ethyl carbonate
2,2-Difluoroethyl ethyl carbonate
Formel:
C₅H₈F₂O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28054292 CAS-Nummer: 916678-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...