4-Amino-3-(trifluoromethyl)phenyl trifluoromethanesulphonate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Amino-3-(trifluoromethyl)phenyl triflate, 2-(Trifluoromethyl)-4-{[(trifluoromethyl)sulphonyl]oxy}aniline
PC48333-1G
PC48333-250MG
APOSPC48333-1GEA
174
EUR
APOSPC48333-1G
APOSPC48333-250MG
4-Amino-3-(trifluoromethyl)phenyl trifluoromethanesulphonate
4-Amino-3-(trifluoromethyl)phenyl trifluoromethanesulphonate
Formel:
C₈H₅F₆NO₃S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD26407433 CAS-Nummer: 1820740-69-9 UN-Nummer: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...