Propen-1-ylsulphur pentafluoride

Lieferant: Apollo Scientific
Achtung

PC450556-1G PC450556-250MG
APOSPC450556-1GEA 574 EUR
APOSPC450556-1G APOSPC450556-250MG
Propen-1-ylsulphur pentafluoride
Propen-1-ylsulphur pentafluoride
Formel: C₃H₅F₅S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31561016
CAS-Nummer: 690-04-0
UN-Nummer: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR