(1-Dimethylamino-2,2,2-trifluoroethoxy)triethylsilane
Lieferant: Apollo Scientific
PC450523-1G
PC450523-5G
APOSPC450523-1GEA
248
EUR
APOSPC450523-1G
APOSPC450523-5G
(1-Dimethylamino-2,2,2-trifluoroethoxy)triethylsilane
(1-Dimethylamino-2,2,2-trifluoroethoxy)triethylsilane
Formel:
C₁₀H₂₂F₃NOSi Siedepunkt: 64…66 °C (5 mm Hg) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD30730306 CAS-Nummer: 208331-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...