Ethyl 2,2,3,3-tetrafluoropropyl carbonate

Lieferant: Apollo Scientific
Achtung

PC450408-1G PC450408-5G
APOSPC450408-1GEA 92.8 EUR
APOSPC450408-1G APOSPC450408-5G
Ethyl 2,2,3,3-tetrafluoropropyl carbonate
Ethyl 2,2,3,3-tetrafluoropropyl carbonate
Formel: C₆H₈F₄O₃
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD28334385
CAS-Nummer: 277332-97-5
UN-Nummer: 3272
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR