6-Chloro-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole
Lieferant: Apollo Scientific
PC450163-1G
PC450163-5G
APOSPC450163-1GEA
237
EUR
APOSPC450163-1G
APOSPC450163-5G
6-Chloro-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole
6-Chloro-5-fluoro-2-(trifluoromethyl)-1H-benzimidazole
Formel:
C₈H₃ClF₄N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01658447 CAS-Nummer: 89426-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...