(3-Fluorophenoxy)acetonitrile 95+%
Lieferant: Apollo Scientific
PC446175-10G
PC446175-1G
PC446175-5G
APOSPC446175-10GEA
248
EUR
APOSPC446175-10G
APOSPC446175-1G
APOSPC446175-5G
(3-Fluorophenoxy)acetonitrile 95+%
(3-Fluorophenoxy)acetonitrile
Formel:
C₈H₆FNO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09928980 CAS-Nummer: 135290-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...