2-(4-Fluorophenyl)acetamide 98%

Lieferant: Apollo Scientific

PC446129-1G PC446129-5G
APOSPC446129-1GEA 72.5 EUR
APOSPC446129-1G APOSPC446129-5G
2-(4-Fluorophenyl)acetamide 98%
2-(4-Fluorophenyl)acetamide
Formel: C₈H₈FNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03576227
CAS-Nummer: 332-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR