5-Aminomethyl-3-(4-fluorophenyl)-[1,2,4]oxadiazole hydrochloride
Lieferant: Apollo Scientific
PC446022-1G
PC446022-5G
APOSPC446022-1GEA
244
EUR
APOSPC446022-1G
APOSPC446022-5G
5-Aminomethyl-3-(4-fluorophenyl)-[1,2,4]oxadiazole hydrochloride
5-Aminomethyl-3-(4-fluorophenyl)-[1,2,4]oxadiazole hydrochloride
Formel:
C₉H₈FN₃O·ClH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12131099 CAS-Nummer: 1208081-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...