7-Fluoro-4-methoxybenzo[d]thiazol-2-amine

Lieferant: Apollo Scientific

PC430539-100MG PC430539-250MG PC430539-1G
APOSPC430539-100MGEA 315 EUR
APOSPC430539-100MG APOSPC430539-250MG APOSPC430539-1G
7-Fluoro-4-methoxybenzo[d]thiazol-2-amine
7-Fluoro-4-methoxybenzo[d]thiazol-2-amine
Formel: C₈H₇FN₂OS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD14585423
CAS-Nummer: 1206969-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR