Potassium trifluoro(3-oxocyclopentyl)borate

Lieferant: Apollo Scientific
Achtung

PC430275-1G PC430275-250MG
APOSPC430275-1GEA 406 EUR
APOSPC430275-1G APOSPC430275-250MG
Potassium trifluoro(3-oxocyclopentyl)borate
Potassium trifluoro(3-oxocyclopentyl)borate
Formel: C₅H₇BF₃O·K
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28348448
CAS-Nummer: 1366170-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR