4-Amino-3,5-difluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-amino-3,5-difluorobenzoicacid

PC430006-1G PC430006-5G PC430006-250MG
APOSPC430006-1GEA 95.7 EUR
APOSPC430006-1G APOSPC430006-5G APOSPC430006-250MG
4-Amino-3,5-difluorobenzoic acid
4-Amino-3,5-difluorobenzoic acid
Formel: C₇H₅F₂NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03407440
CAS-Nummer: 500577-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR