4-Chloro-5-(difluoromethyl)-2-fluoro-1,3-thiazole 98%

Lieferant: Apollo Scientific
Achtung

PC4170-1G PC4170-5G
APOSPC4170-1GEA 357 EUR
APOSPC4170-1G APOSPC4170-5G
4-Chloro-5-(difluoromethyl)-2-fluoro-1,3-thiazole 98%
4-Chloro-5-(difluoromethyl)-2-fluoro-1,3-thiazole
Formel: C₄HClF₃NS
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD04972713
CAS-Nummer: 105315-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR