2-Chloro-4-fluoro-5-methoxyphenylboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Chloro-4-fluoro-5-methoxyphenyl)boronic acid
PC412302-1G
PC412302-250MG
APOSPC412302-1GEA
284
EUR
APOSPC412302-1G
APOSPC412302-250MG
2-Chloro-4-fluoro-5-methoxyphenylboronic acid 98%
2-Chloro-4-fluoro-5-methoxyphenylboronic acid
Formel:
C₇H₇BClFO₃ Molekulargewicht: 204.391 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09998845 CAS-Nummer: 1256355-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...