4-(3,5-Difluorophenoxy)phenylboronic acid
Lieferant: Apollo Scientific
Synonyme:
(4-(3,5-Difluorophenoxy)phenyl)boronic acid
PC412134-1G
PC412134-250MG
APOSPC412134-1GEA
453
EUR
APOSPC412134-1G
APOSPC412134-250MG
4-(3,5-Difluorophenoxy)phenylboronic acid
4-(3,5-Difluorophenoxy)phenylboronic acid
Formel:
C₁₂H₉BF₂O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD14636486 CAS-Nummer: 1029438-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...