2,2,2-Trifluoro-1-pyridin-2-ylethanamine dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
PC411329-1G
PC411329-5G
APOSPC411329-1GEA
284
EUR
APOSPC411329-1G
APOSPC411329-5G
2,2,2-Trifluoro-1-pyridin-2-ylethanamine dihydrochloride
2,2,2-Trifluoro-1-pyridin-2-ylethanamine dihydrochloride
Formel:
C₇H₇F₃N₂·2ClH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16872039 CAS-Nummer: 1241675-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...