4-Methyl-2-(2,2,2-trifluoroethoxy)aniline
Lieferant: Apollo Scientific
|
Achtung
|
PC411313-1G
PC411313-5G
APOSPC411313-1GEA
69.6
EUR
APOSPC411313-1G
APOSPC411313-5G
4-Methyl-2-(2,2,2-trifluoroethoxy)aniline
4-Methyl-2-(2,2,2-trifluoroethoxy)aniline
Formel:
C₉H₁₀F₃NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09943424 CAS-Nummer: 186387-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...