cis-3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride

Lieferant: Apollo Scientific

PC411279-100MG PC411279-1G PC411279-250MG
APOSPC411279-100MGEA 432 EUR
APOSPC411279-100MG APOSPC411279-1G APOSPC411279-250MG
cis-3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride
cis-3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride
Formel: C₅H₈F₃NO·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD23106129
CAS-Nummer: 1408075-93-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR