4-Ethyl-5-fluoro-2-methoxyphenol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Ethyl-5-fluoro-o-guaiacol, 5-Ethyl-4-fluoro-2-hydroxyanisole

PC4081-1G PC4081-5G
APOSPC4081-1GEA 105 EUR
APOSPC4081-1G APOSPC4081-5G
4-Ethyl-5-fluoro-2-methoxyphenol
4-Ethyl-5-fluoro-2-methoxyphenol
Formel: C₉H₁₁FO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11101442
CAS-Nummer: 1065076-45-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR